AMBH303C6205 Ambeed, Inc.

Ethyl 2-bromo-3-methylbutanoate

  • MDL number MFCD00026855
  • Cas Number : 609-12-1
  • Molecular Weight : 209.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/609-12-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/609-12-1.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart