AMBH303C6223 Ambeed, Inc.

2,6-Dichloro-4-nitrophenol

  • MDL number MFCD00014715
  • Cas Number : 618-80-4
  • Molecular Weight : 208.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/618-80-4.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P311
MSDS https://www.ambeed.com/products/618-80-4.html
pictograms GHS06
hstatements H302+H312-H315-H319-H331-H335
Availability details for
Quantity
  • Availability
  • Add to Cart