AMBH303C622E Ambeed, Inc.

N-(3-(2-(tert-Butyl)-5-(2-chloropyrimidin-4-yl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulfonamide

  • MDL number MFCD18447700
  • Cas Number : 1195768-23-0
  • Molecular Weight : 538.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1195768-23-0.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1195768-23-0.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart