AMBH303C626B Ambeed, Inc.

Aminophylline

  • MDL number MFCD00013221
  • Cas Number : 317-34-0
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/317-34-0.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/317-34-0.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart