AMBH303C627C Ambeed, Inc.

6-Ethylpyridin-3-amine

  • MDL number MFCD11111131
  • Cas Number : 126553-00-2
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 125
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 244�t 760 mmHg
Product Link https://www.ambeed.com/products/126553-00-2.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 6.1
MSDS https://www.ambeed.com/products/126553-00-2.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart