AMBH303C6289 Ambeed, Inc.

2-((3-Cyano-4-(4-methoxyphenyl)-6-phenylpyridin-2-yl)thio)-N,N-dicyclohexylacetamide

  • MDL number MFCD01817480
  • Cas Number : 332101-03-8
  • Molecular Weight : 539.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/332101-03-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/332101-03-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart