AMBH303C6296 Ambeed, Inc.

2-Butylthiophene

  • MDL number MFCD00022534
  • Cas Number : 1455-20-5
  • Molecular Weight : 140.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 56
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 181�t 760 mmHg
Product Link https://www.ambeed.com/products/1455-20-5.html

Safety Information

signalword Danger
pcodes P233-P242-P280-P370+P378-P403+P235-P501
hcodes 3
MSDS https://www.ambeed.com/products/1455-20-5.html
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart