AMBH303C62A0 Ambeed, Inc.

Palomid 529

  • MDL number MFCD18633224
  • Cas Number : 914913-88-5
  • Molecular Weight : 406.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-(1-Hydroxyethyl)-2-methoxy-3-((4-methoxybenzyl)oxy)-6H-benzo[c]chromen-6-one
Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/914913-88-5.html

Safety Information

signalword Warning
pcodes P261-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/914913-88-5.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart