AMBH303C62D9 Ambeed, Inc.

2-Chloro-3-(methoxymethyl)pyrazine

  • MDL number MFCD18837167
  • Cas Number : 1289387-97-8
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1289387-97-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1289387-97-8.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart