AMBH303C6301 Ambeed, Inc.

Pioglitazone

  • MDL number MFCD00865504
  • Cas Number : 111025-46-8
  • Molecular Weight : 356.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/111025-46-8.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/111025-46-8.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart