AMBH303C630E Ambeed, Inc.

3-Methyl-4-nitroaniline

  • MDL number MFCD00091833
  • Cas Number : 611-05-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/611-05-2.html

Safety Information

signalword Danger
pcodes P280-P273-P309-P310-P302+P352
MSDS https://www.ambeed.com/products/611-05-2.html
pictograms GHS06,GHS08,GHS09
hstatements H301+H311+H331-H373-H401-H411
Availability details for
Quantity
  • Availability
  • Add to Cart