AMBH303C6311 Ambeed, Inc.

N-Methyltetrahydrofuran-3-amine hydrochloride

  • MDL number MFCD09952761
  • Cas Number : 917882-94-1
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/917882-94-1.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/917882-94-1.html
pictograms GHS05
hstatements H318
Availability details for
Quantity
  • Availability
  • Add to Cart