AMBH303C632C Ambeed, Inc.

(Z)-N-(1-Chloro-1-(4-methoxyphenyl)-3-oxo-3-(piperidin-1-yl)prop-1-en-2-yl)benzamide

  • MDL number MFCD29037392
  • Cas Number : 1323140-64-2
  • Molecular Weight : 398.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 338.3
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 635.7�t 760 mmHg
Product Link https://www.ambeed.com/products/1323140-64-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1323140-64-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart