AMBH303C6332 Ambeed, Inc.

4-(3-Iodo-1H-pyrazol-4-yl)-2-(methylthio)pyrimidine

  • MDL number MFCD12756165
  • Cas Number : 1111638-74-4
  • Molecular Weight : 318.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1111638-74-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1111638-74-4.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart