AMBH303C633F Ambeed, Inc.

4-Amino-1-(pyridin-3-yl)butan-1-one dihydrochloride

  • MDL number MFCD03425485
  • Cas Number : 1187930-07-9
  • Molecular Weight : 237.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1187930-07-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1187930-07-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart