AMBH303C6362 Ambeed, Inc.

Methyl 2-(2-methoxypyridin-4-yl)acetate

  • MDL number MFCD22572382
  • Cas Number : 464152-37-2
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/464152-37-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/464152-37-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart