AMBH303C636E Ambeed, Inc.

2-(tert-Butyl)-6-nitro-1H-indole

  • MDL number MFCD22689954
  • Cas Number : 873055-09-5
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/873055-09-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/873055-09-5.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart