AMBH303C6373 Ambeed, Inc.

1-((2-Hydroxyethyl)amino)propan-2-ol

  • MDL number MFCD00021892
  • Cas Number : 6579-55-1
  • Molecular Weight : 119.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6579-55-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/6579-55-1.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart