AMBH303C637D Ambeed, Inc.

Methyl 5-(aminomethyl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD13174856
  • Cas Number : 263382-27-0
  • Molecular Weight : 154.1700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/263382-27-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/263382-27-0.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart