AMBH303C63B4 Ambeed, Inc.

Methyl 2-(pyrimidin-4-yl)acetate

  • MDL number MFCD13705337
  • Cas Number : 863032-29-5
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/863032-29-5.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/863032-29-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart