AMBH303C63D3 Ambeed, Inc.

1-(4-Iodophenyl)ethanamine

  • MDL number MFCD05215387
  • Cas Number : 90086-41-2
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Yellow to Brown Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/90086-41-2.html

Safety Information

signalword Warning
pcodes P261-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/90086-41-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart