AMBH303C63E1 Ambeed, Inc.

3,4-Dimethoxybenzyl Chloride

  • MDL number MFCD00185554
  • Cas Number : 7306-46-9
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-53
Purity 95
Flash Point 98.6
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 260.8�t 760 mmHg
Product Link https://www.ambeed.com/products/7306-46-9.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/7306-46-9.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart