AMBH303C63F9 Ambeed, Inc.

5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one

  • MDL number MFCD30729989
  • Cas Number : 164391-52-0
  • Molecular Weight : 350.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/164391-52-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/164391-52-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart