AMBH303C640F Ambeed, Inc.

4-Amino-1H-imidazole-2-carboxylic acid

  • MDL number MFCD00984820
  • Cas Number : 155815-92-2
  • Molecular Weight : 127.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/155815-92-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/155815-92-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart