AMBH303C6414 Ambeed, Inc.

4-(2-Bromopyridin-4-yl)morpholine

  • MDL number MFCD11869654
  • Cas Number : 1049023-41-7
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 372.2�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1049023-41-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1049023-41-7.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart