AMBH303C641E Ambeed, Inc.

Stannous chloride dihydrate

  • MDL number MFCD00149863
  • Cas Number : 10025-69-1
  • Molecular Weight : 225.6466

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/10025-69-1.html

Safety Information

signalword Danger
pcodes P260-P273-P280-P303+P361+P353-P304+P340+P310-P305+P351+P338-P391
MSDS https://www.ambeed.com/products/10025-69-1.html
pictograms GHS05,GHS07,GHS08,GHS09
hstatements H302+H332-H314-H317-H373-H410
Availability details for
Quantity
  • Availability
  • Add to Cart