AMBH303C6424 Ambeed, Inc.

Dimethyl 3-bromopyridine-2,4-dicarboxylate

  • Cas Number : 1222185-12-7
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1222185-12-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1222185-12-7.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart