AMBH303C6427 Ambeed, Inc.

2-(2-Fluorophenyl)piperidine hydrochloride

  • MDL number MFCD02178959
  • Cas Number : 1185010-62-1
  • Molecular Weight : 215.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1185010-62-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1185010-62-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart