AMBH303C642D Ambeed, Inc.

tert-Butyl 3-((2-methoxyethyl)amino)pyrrolidine-1-carboxylate

  • MDL number MFCD06656605
  • Cas Number : 887587-33-9
  • Molecular Weight : 244.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/887587-33-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/887587-33-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart