AMBH303C6438 Ambeed, Inc.

1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethanone

  • MDL number MFCD09033719
  • Cas Number : 13577-40-7
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13577-40-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13577-40-7.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart