AMBH303C643D Ambeed, Inc.

4-Chloro-6-methyl-2-trifluoromethylpyrimidine

  • MDL number MFCD09743686
  • Cas Number : 1582-25-8
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 147.2�t 760 mmHg
Product Link https://www.ambeed.com/products/1582-25-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1582-25-8.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart