AMBH303C645E Ambeed, Inc.

(R)-tert-Butyl 2-benzylpiperazine-1-carboxylate

  • MDL number MFCD07772094
  • Cas Number : 947684-78-8
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/947684-78-8.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/947684-78-8.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart