AMBH303C6469 Ambeed, Inc.

SKL2001

  • MDL number N/A
  • Cas Number : 909089-13-0
  • Molecular Weight : 286.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(3-(1H-Imidazol-1-yl)propyl)-5-(furan-2-yl)isoxazole-3-carboxamide
Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/909089-13-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/909089-13-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart