AMBH303C648B Ambeed, Inc.

2,5-Dibromo-3,6-dimethylpyridine

  • MDL number MFCD00234281
  • Cas Number : 38749-93-8
  • Molecular Weight : 264.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/38749-93-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/38749-93-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart