AMBH303C64A5 Ambeed, Inc.

2,4-Dimethoxy-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number MFCD12406107
  • Cas Number : 84538-40-9
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/84538-40-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/84538-40-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart