AMBH303C64AD Ambeed, Inc.

6,7-Dimethoxy-2-methyl-4-phenyl-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD11501291
  • Cas Number : 128942-65-4
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-81
Purity 95
Flash Point 131.1
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 393.4�0�t 760 mmHg
Product Link https://www.ambeed.com/products/128942-65-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/128942-65-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart