AMBH303C64B1 Ambeed, Inc.

Cupric bromide

  • MDL number MFCD00010970
  • Cas Number : 7789-45-9
  • Molecular Weight : 223.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/7789-45-9.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/7789-45-9.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart