AMBH303C64F4 Ambeed, Inc.

Ceftezole sodium

  • MDL number MFCD01682031
  • Cas Number : 41136-22-5
  • Molecular Weight : 462.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium (6R,7R)-3-(((1,3,4-thiadiazol-2-yl)thio)methyl)-7-(2-(1H-tetrazol-1-yl)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Purity 90
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/41136-22-5.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/41136-22-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart