AMBH303C652F Ambeed, Inc.

2-Bromophenylhydrazine hydrochloride

  • MDL number MFCD00012926
  • Cas Number : 50709-33-6
  • Molecular Weight : 223.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/50709-33-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/50709-33-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart