AMBH303C6531 Ambeed, Inc.

Ethyl 1-oxaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD12498695
  • Cas Number : 171361-65-2
  • Molecular Weight : 184.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/171361-65-2.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/171361-65-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart