AMBH303C6549 Ambeed, Inc.

1-(Tert-butoxycarbonyl)-5,5-dimethylpyrrolidine-2-carboxylic acid

  • MDL number MFCD13190739
  • Cas Number : 900158-99-8
  • Molecular Weight : 243.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/900158-99-8.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/900158-99-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart