AMBH303C6590 Ambeed, Inc.

1-((1S,2S)-1-Hydroxy-1-(4-hydroxyphenyl)propan-2-yl)-4-phenylpiperidin-4-ol methanesulfonate trihydrate

  • MDL number MFCD22377951
  • Cas Number : 189894-57-3
  • Molecular Weight : 477.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/189894-57-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/189894-57-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart