AMBH303C6594 Ambeed, Inc.

6-Chloro-2-ethylpyrimidin-4-amine

  • MDL number MFCD11045678
  • Cas Number : 98134-36-2
  • Molecular Weight : 157.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/98134-36-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/98134-36-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart