AMBH303C6596 Ambeed, Inc.

Methyl 5-fluoro-2,6-dihydroxynicotinate

  • MDL number MFCD27996650
  • Cas Number : 148874-68-4
  • Molecular Weight : 187.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/148874-68-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/148874-68-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart