AMBH303C659A Ambeed, Inc.

YU238259

  • MDL number MFCD30532769
  • Cas Number : 1943733-16-1
  • Molecular Weight : 459.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(2-(5-Chloropyridin-2-yl)ethyl)-4-((4-methoxyphenylsulfonamido)methyl)benzamide
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1943733-16-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1943733-16-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart