AMBH303C6606 Ambeed, Inc.

5-Methylhexane-2,3-dione

  • MDL number MFCD00059352
  • Cas Number : 13706-86-0
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13706-86-0.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13706-86-0.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart