AMBH303C662C Ambeed, Inc.

1,2-Dibromo-4-fluoro-5-methylbenzene

  • MDL number MFCD00143234
  • Cas Number : 202982-77-2
  • Molecular Weight : 267.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/202982-77-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/202982-77-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart