AMBH303C663A Ambeed, Inc.

N-((4-(4-Fluorobenzyl)morpholin-2-yl)methyl)acetamide

  • MDL number MFCD16619692
  • Cas Number : 112913-94-7
  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172
Purity 95
Flash Point 233.7
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 462.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/112913-94-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112913-94-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart