AMBH303C6661 Ambeed, Inc.

2,2-Dihydroxy-1-(4-hydroxyphenyl)ethanone

  • MDL number MFCD14706138
  • Cas Number : 197447-05-5
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/197447-05-5.html

Safety Information

signalword Warning
pcodes P280-P301+P310-P261
MSDS https://www.ambeed.com/products/197447-05-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart