AMBH303C6672 Ambeed, Inc.

2-(5-Nitro-1H-indazol-1-yl)ethanol

  • MDL number MFCD14708234
  • Cas Number : 1056619-14-7
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1056619-14-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1056619-14-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart